Assay Details
Gene Symbol: Rad23a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001297606.1 | NP_001284535.1 | 4 - 5 | 576 | 67 | |
NM_001297607.1 | NP_001284536.1 | 4 - 5 | 576 | 67 | ||
NM_009010.5 | NP_033036.2 | 4 - 5 | 576 | 67 | ||
GenBank mRNA | AK038300.1 | - | 4 - 5 | 553 | 67 | |
AK045224.1 | - | 4 - 5 | 566 | 67 | ||
AK161875.1 | - | 4 - 5 | 553 | 67 | ||
AK165467.1 | - | 4 - 5 | 577 | 67 | ||
BC003846.1 | - | 4 - 5 | 564 | 67 | ||
BC138684.1 | - | 4 - 5 | 536 | 8861705 | 67 | |
BC145372.1 | - | 4 - 5 | 536 | 9053930 | 67 | |
X92410.1 | - | 4 - 5 | 476 | 67 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
