Assay Details
Gene Symbol: Rad23a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001297606.1 | NP_001284535.1 | 6 - 7 | 782 | 88 | |
NM_001297607.1 | NP_001284536.1 | 6 - 7 | 782 | 88 | ||
GenBank mRNA | AK161875.1 | - | 6 - 7 | 759 | 88 | |
AK165467.1 | - | 6 - 7 | 783 | 88 | ||
BC003846.1 | - | 6 - 7 | 770 | 88 | ||
BC145372.1 | - | 6 - 7 | 742 | 9053930 | 88 | |
X92410.1 | - | 6 - 7 | 682 | 88 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
Function(s)
Process(es)
DNA repair
nucleotide-excision repair
cellular response to DNA damage stimulus
regulation of proteasomal ubiquitin-dependent protein catabolic process
proteasome-mediated ubiquitin-dependent protein catabolic process
positive regulation of viral genome replication
damaged DNA binding
polyubiquitin modification-dependent protein binding
ubiquitin-specific protease binding
DNA repair
nucleotide-excision repair
cellular response to DNA damage stimulus
regulation of proteasomal ubiquitin-dependent protein catabolic process
proteasome-mediated ubiquitin-dependent protein catabolic process
positive regulation of viral genome replication
damaged DNA binding
polyubiquitin modification-dependent protein binding
ubiquitin-specific protease binding
MicroArray Probes:
Agilent
Affymetrix
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P26022217 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P26022222 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P26022218 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P26022219 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P26022220 | - | - |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1985749 | AK028227.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1985749 | NM_183358.4 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1985749 | NM_009010.4 | mouse_transcriptome |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 488503 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 427559 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 559710 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 119132 | - | - |
Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P179210 | - | - |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1985749 | AK028227.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1985749 | NM_183358.4 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1985749 | NM_009010.4 | mouse_transcriptome |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26022204 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26022224 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26022231 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26022228 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26022205 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26022229 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26022211 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26022207 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26022214 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26022227 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26022220 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26022216 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26022219 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P06579320 | - | - |
Whole Mouse Genome G4122A | A_52_P48206 | BC003846.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P48206 | BC138684.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P48206 | BC145372.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P48206 | X92410.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P48206 | AK080645.1 | mouse_transcriptome |
Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P131972 | - | - |
Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P108801 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P07189875 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P07189876 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P07189885 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P07189869 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P26022222 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P26022219 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P26022217 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P26022220 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P26022218 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26022174 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26022178 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26022278 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26022220 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26022287 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26022160 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26022179 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26022217 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26022268 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26022272 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26022224 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26022182 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26022184 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26022252 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26022159 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26022227 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26022211 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26022210 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26022258 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26022207 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26022280 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26022165 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26022266 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26022166 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26022163 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26022186 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26022229 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26022169 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26022282 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26022199 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26022168 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26022202 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26022181 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26022185 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26022195 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26022260 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26022273 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26022204 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26022233 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26022277 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26022188 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26022164 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26022205 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26022228 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26022200 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26022271 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26022192 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26022283 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26022264 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26022161 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26022255 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26022176 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26022263 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26022269 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26022172 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26022156 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26022152 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26022276 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26022153 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26022231 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26022191 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26022151 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26022197 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26022281 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26022170 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26022146 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26022158 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26022190 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26022214 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26022143 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26022257 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26022184 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26022217 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P07189880 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P07213070 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26022233 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26022195 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26022191 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26022200 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P06829785 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26022188 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26022207 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26022199 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26022228 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26022197 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P06829786 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26022190 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26022192 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P07133339 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P06711872 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26022227 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26022204 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26022224 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26022214 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26022220 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P07133324 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P07189886 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26022198 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P07189876 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26022202 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Mouse Gene 1.0 ST Array | 10580214 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10580213 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10580212 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10580218 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10573433 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10580215 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10580211 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10580216 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10580217 | - | - |
Affymetrix HT MG-430 PM Array Plate | 1453623_PM_a_at | - | - |
Affymetrix HT MG-430 PM Array Plate | 1455391_PM_at | - | - |
Affymetrix HT MG-430 PM Array Plate | 1437697_PM_at | - | - |
Affymetrix HT MG-430 PM Array Plate | 1422964_PM_at | - | - |
Affymetrix Mouse Gene 1.1 ST Array | 10580210 | - | - |
Affymetrix Mouse Gene 1.1 ST Array | 10573430 | - | - |
Mouse Genome 430 2.0 Array | 1446783_PM_at | - | - |
Mouse Genome 430 2.0 Array | 1455391_at | - | - |
Mouse Genome 430 2.0 Array | 1455391_PM_at | - | - |
Mouse Genome 430 2.0 Array | 1422964_at | - | - |
Mouse Genome 430 2.0 Array | 1446783_at | - | - |
Mouse Genome 430 2.0 Array | 1453623_a_at | - | - |
Mouse Genome 430 2.0 Array | 1437697_PM_at | - | - |
Mouse Genome 430 2.0 Array | 1437697_at | - | - |
Mouse Genome 430 2.0 Array | 1422964_PM_at | - | - |
Mouse Genome 430 2.0 Array | 1453623_PM_a_at | - | - |
Affymetrix GeneChip Mouse Genome 430 2.0 Array | 19358_at | - | - |
Affymetrix GeneChip Mouse Genome 430 2.0 Array | 19358_PM_at | - | - |
Affymetrix Mouse Clariom S Array | PSR0800019786.mm.1 | TC0800002592.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR0800019786.mm.1 | NM_009010.5 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0800019784.mm.1 | TC0800002592.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR0800019784.mm.1 | NM_009010.5 | Refseq |
Mouse Expression Set 430 | 1437697_PM_at | - | - |
Mouse Expression Set 430 | 1422964_PM_at | - | - |
Mouse Expression Set 430 | 1422964_at | - | - |
Mouse Expression Set 430 | 1437697_at | - | - |
Mouse Expression Set 430 | 1455391_at | - | - |
Mouse Expression Set 430 | 1455391_PM_at | - | - |
Murine Genome U74v2 Set | 92410_PM_at | - | - |
Murine Genome U74v2 Set | 92410_at | - | - |
Mouse Genome 430A 2.0 Array | 1422964_at | - | - |
Mouse Genome 430A 2.0 Array | 1437697_PM_at | - | - |
Mouse Genome 430A 2.0 Array | 1437697_at | - | - |
Mouse Genome 430A 2.0 Array | 1422964_PM_at | - | - |
Affymetrix HT Mouse Genome 430A Array | 1422964_at | - | - |
Affymetrix HT Mouse Genome 430A Array | 1453623_a_at | - | - |
Affymetrix HT Mouse Genome 430A Array | 1422964_PM_at | - | - |
Affymetrix HT Mouse Genome 430A Array | 1437697_at | - | - |
Affymetrix HT Mouse Genome 430A Array | 1437697_PM_at | - | - |
Affymetrix HT Mouse Genome 430A Array | 1453623_PM_a_at | - | - |
Affymetrix HT Mouse Genome 430B Array | 1455391_at | - | - |
Affymetrix HT Mouse Genome 430B Array | 1455391_PM_at | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
MouseWG-6 v2 Expression BeadChip | 5270309 | - | - |
MouseWG-6 v2 Expression BeadChip | 5360463 | - | - |
MouseWG-6 v2 Expression BeadChip | 5550364 | - | - |
Illumina mouse-6 v1.0 expression beadchip | 5270309 | - | - |
Illumina mouse-6 v1.0 expression beadchip | 5360463 | - | - |
Illumina mouse-6 v1.0 expression beadchip | 5550364 | - | - |
Illumina mouse-6 v1.0 expression beadchip | 670082 | - | - |
Illumina mouseRef-8 v1.0 expression beadchip | 0000670082 | - | - |
Illumina mouseRef-8 v1.0 expression beadchip | 0005270309 | - | - |
Illumina mouseRef-8 v1.0 expression beadchip | 0005550364 | - | - |
Illumina mouseRef-8 v1.0 expression beadchip | 0005360463 | - | - |
Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2745692 | - | - |
Illumina mouseRef-8 v1.1 expression beadchip | ILMN_1240677 | - | - |
Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2662743 | - | - |
Illumina mouseRef-8 v1.1 expression beadchip | ILMN_1216295 | - | - |
Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2711810 | - | - |
Illumina mouse-6 v1.1 expression beadchip | ILMN_2662743 | - | - |
Illumina mouse-6 v1.1 expression beadchip | ILMN_2711810 | - | - |
Illumina mouse-6 v1.1 expression beadchip | ILMN_1240677 | - | - |
Illumina mouse-6 v1.1 expression beadchip | ILMN_2745692 | - | - |
Illumina mouse-6 v1.1 expression beadchip | ILMN_1216295 | - | - |
mouse-6 v1.1 (Illumina) | scl0019358.2_167-S | - | - |
mouse-6 v1.1 (Illumina) | scl34556.8.1_48-S | - | - |
mouse-6 v1.1 (Illumina) | scl019358.2_23-S | - | - |
mouse-6 v1.1 (Illumina) | scl000680.1_0-S | - | - |
Illumina Sentrix mouse-6.v1 bead array | 5550364 | - | - |
Illumina Sentrix mouse-6.v1 bead array | 5360463 | - | - |
Illumina Sentrix mouse-6.v1 bead array | 5270309 | - | - |
Illumina Sentrix mouse-6.v1 bead array | 670082 | - | - |
Illumina Sentrix MouseRef-8 Expression BeadChip | scl019358.2_23-S | - | - |
Illumina Sentrix MouseRef-8 Expression BeadChip | scl34556.8.1_48-S | - | - |
Illumina Sentrix MouseRef-8 Expression BeadChip | scl000680.1_0-S | - | - |
Illumina Sentrix MouseRef-8 Expression BeadChip | scl0019358.2_167-S | - | - |
Illumina Sentrix Mouse-6 Expression BeadChip (2) | 5360463 | - | - |
Illumina Sentrix Mouse-6 Expression BeadChip (2) | 5550364 | - | - |
Illumina Sentrix Mouse-6 Expression BeadChip (2) | 5270309 | - | - |
Illumina Sentrix Mouse-6 Expression BeadChip (2) | 670082 | - | - |
MouseRef-8 v2 Expression BeadChip | ILMN_1216295 | - | - |
MouseRef-8 v2 Expression BeadChip | ILMN_2662743 | - | - |
MouseRef-8 v2 Expression BeadChip | ILMN_2637052 | - | - |
Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2745692 | - | - |
Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2637052 | - | - |
Illumina MouseWG-6 v2.0 expression beadchip | ILMN_1216295 | - | - |
Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2662743 | - | - |
Illumina Sentrix Mouse-6 Expression BeadChip (3) | scl019358.2_23-S | - | - |
