Assay Details
Gene Symbol: Ralgds | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001145834.1 | NP_001139306.1 | 4 - 5 | 754 | 78 | |
NM_001145835.1 | NP_001139307.1 | 4 - 5 | 666 | 78 | ||
NM_001145836.1 | NP_001139308.1 | 4 - 5 | 630 | 78 | ||
NM_009058.2 | NP_033084.2 | 4 - 5 | 790 | 78 | ||
GenBank mRNA | AK129328.1 | - | 4 - 5 | 631 | 78 | |
AK141959.1 | - | 4 - 5 | 621 | 78 | ||
AK159020.1 | - | 4 - 5 | 623 | 78 | ||
AK159754.1 | - | 4 - 5 | 636 | 78 | ||
BC027225.1 | - | 4 - 5 | 448 | 3486732 | 78 | |
L07924.1 | - | 4 - 5 | 626 | 78 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
