Assay Details
Gene Symbol: Ralgds | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001145834.1 | NP_001139306.1 | 16 - 17 | 2610 | 61 | |
NM_001145835.1 | NP_001139307.1 | 16 - 17 | 2522 | 61 | ||
NM_001145836.1 | NP_001139308.1 | 16 - 17 | 2486 | 61 | ||
NM_009058.2 | NP_033084.2 | 16 - 17 | 2646 | 61 | ||
GenBank mRNA | AK129328.1 | - | 16 - 17 | 2487 | 61 | |
AK141959.1 | - | 16 - 17 | 2477 | 61 | ||
AK149644.1 | - | 11 - 12 | 1381 | 61 | ||
AK159020.1 | - | 16 - 17 | 2479 | 61 | ||
AK159754.1 | - | 16 - 17 | 2492 | 61 | ||
AK188792.1 | - | 2 - 3 | 183 | 61 | ||
BC027225.1 | - | 16 - 17 | 2304 | 3486732 | 61 | |
L07924.1 | - | 16 - 17 | 2482 | 61 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
