Assay Details
Gene Symbol: Rgl2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_009059.2 | NP_033085.2 | 7 - 8 | 1217 | 76 | |
GenBank mRNA | BC068121.1 | - | 7 - 8 | 1217 | 6415928 | 76 |
KJ650210.1 | - | 6 - 7 | 1025 | 76 | ||
U54639.1 | - | 7 - 8 | 1146 | 76 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
