Assay Details
Gene Symbol: Rgl2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_009059.2 | NP_033085.2 | 16 - 17 | 2198 | 134 | |
GenBank mRNA | AK144440.1 | - | 16 - 17 | 2201 | 134 | |
BC068121.1 | - | 16 - 17 | 2198 | 6415928 | 134 | |
KJ650210.1 | - | 15 - 16 | 2006 | 134 | ||
U54639.1 | - | 16 - 17 | 2127 | 134 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
