Assay Details
Gene Symbol: Rps6ka1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001285505.1 | NP_001272434.1 | 14 - 15 | 1351 | 78 | |
NM_001285506.1 | NP_001272435.1 | 13 - 14 | 1235 | 78 | ||
NM_009097.5 | NP_033123.2 | 14 - 15 | 1384 | 78 | ||
GenBank mRNA | AK132856.1 | - | 12 - 13 | 1421 | 78 | |
AK148208.1 | - | 14 - 15 | 1377 | 78 | ||
AK160571.1 | - | 2 - 3 | 184 | 78 | ||
AK167790.1 | - | 13 - 14 | 1222 | 78 | ||
BC049076.1 | - | 14 - 15 | 1335 | 78 | ||
BC094470.1 | - | 14 - 15 | 1337 | 5098650 | 78 | |
M28489.1 | - | 14 - 15 | 1337 | 78 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
