Assay Details
Gene Symbol: Ulk1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_009469.3 | NP_033495.2 | 24 - 25 | 3041 | 92 | |
GenBank mRNA | AF053756.1 | - | 24 - 25 | 2733 | 92 | |
AF072370.1 | - | 24 - 25 | 2724 | 92 | ||
AK122357.1 | - | 23 - 24 | 2543 | 92 | ||
AK148317.1 | - | 24 - 25 | 3034 | 92 | ||
AK154939.1 | - | 24 - 25 | 3058 | 92 | ||
AK156534.1 | - | 24 - 25 | 3018 | 92 | ||
BC030850.1 | - | 3 - 4 | 513 | 92 | ||
BC053062.1 | - | 24 - 25 | 2701 | 92 | ||
BC057121.1 | - | 24 - 25 | 3041 | 6834534 | 92 | |
BC059835.1 | - | 24 - 25 | 3010 | 6406755 | 92 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
