Assay Details
Gene Symbol: Chrne | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_009603.1 | NP_033733.1 | 3 - 4 | 259 | 70 | |
GenBank mRNA | AF032863.1 | - | 2 - 3 | 112 | 70 | |
X55718.1 | - | 3 - 4 | 259 | 70 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
