Assay Details
Gene Symbol: Eda | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001177937.1 | NP_001171408.1 | 4 - 5 | 880 | 81 | |
NM_001177938.1 | NP_001171409.1 | 4 - 5 | 880 | 81 | ||
NM_001177939.1 | NP_001171410.1 | 4 - 5 | 880 | 81 | ||
NM_001177940.1 | NP_001171411.1 | 3 - 4 | 856 | 81 | ||
NM_001177941.1 | NP_001171412.1 | 3 - 4 | 856 | 81 | ||
NM_001177942.1 | NP_001171413.1 | 4 - 5 | 880 | 81 | ||
NM_001177943.1 | NP_001171414.1 | 4 - 5 | 880 | 81 | ||
NM_001177944.1 | NP_001171415.1 | 4 - 5 | 880 | 81 | ||
NM_010099.2 | NP_034229.1 | 4 - 5 | 880 | 81 | ||
GenBank mRNA | AF004435.1 | - | 4 - 5 | 846 | 81 | |
AF016628.1 | - | 4 - 5 | 880 | 81 | ||
AF016629.1 | - | 4 - 5 | 880 | 81 | ||
AJ243657.1 | - | 4 - 5 | 867 | 81 | ||
AJ243658.1 | - | 4 - 5 | 828 | 81 | ||
AK137551.1 | - | 4 - 5 | 789 | 81 | ||
BC125387.1 | - | 4 - 5 | 771 | 40129902 | 81 | |
BC144790.1 | - | 4 - 5 | 771 | 9053338 | 81 | |
BC144791.1 | - | 4 - 5 | 771 | 9053339 | 81 | |
DQ168876.1 | - | 4 - 5 | 880 | 81 | ||
DQ168877.1 | - | 4 - 5 | 880 | 81 | ||
DQ168878.1 | - | 3 - 4 | 856 | 81 | ||
DQ168879.1 | - | 3 - 4 | 856 | 81 | ||
DQ168880.1 | - | 4 - 5 | 880 | 81 | ||
DQ168881.1 | - | 4 - 5 | 880 | 81 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
