Assay Details
Gene Symbol: Irs1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_010570.4 | NP_034700.2 | 1 | 2597 | 113 | |
GenBank mRNA | AK045317.1 | - | 1 | 2575 | 113 | |
AK142242.1 | - | 1 | 2064 | 113 | ||
L24563.1 | - | 1 | 1651 | 113 | ||
X69722.1 | - | 1 | 2123 | 113 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
