Assay Details
Gene Symbol: Itga2b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_010575.2 | NP_034705.2 | 8 - 9 | 1010 | 75 | |
GenBank mRNA | AF166384.1 | - | 8 - 9 | 888 | 75 | |
AF170316.1 | - | 8 - 9 | 871 | 75 | ||
AK048700.1 | - | 4 - 5 | 365 | 75 | ||
AK140749.1 | - | 3 - 4 | 662 | 75 | ||
AK142289.1 | - | 8 - 9 | 872 | 75 | ||
AK154619.1 | - | 8 - 9 | 1011 | 75 | ||
AK156872.1 | - | 7 - 8 | 1192 | 75 | ||
AK157012.1 | - | 7 - 8 | 1192 | 75 | ||
AK158443.1 | - | 8 - 9 | 857 | 75 | ||
BC120493.1 | - | 8 - 9 | 860 | 40129416 | 75 | |
BC137570.1 | - | 8 - 9 | 860 | 8860589 | 75 |

More Information
Updated Mapping Information

Additional Information:
This assay is in the 5'UTR region of the AK048700.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
