Assay Details
Gene Symbol: Itk | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001281965.1 | NP_001268894.1 | 11 - 12 | 1179 | 94 | |
NM_001281966.1 | NP_001268895.1 | 11 - 12 | 1170 | 94 | ||
NM_001281968.1 | NP_001268897.1 | 11 - 12 | 1158 | 94 | ||
NM_010583.3 | NP_034713.2 | 11 - 12 | 1161 | 94 | ||
GenBank mRNA | AK037554.1 | - | 241 | 94 | ||
AK083380.1 | - | 11 - 12 | 1133 | 94 | ||
AK138113.1 | - | 3 - 4 | 95 | |||
AK153613.1 | - | 11 - 12 | 1117 | 94 | ||
BC128374.1 | - | 11 - 12 | 1087 | 40111687 | 94 | |
BC128375.1 | - | 11 - 12 | 1084 | 40111688 | 94 | |
D14042.1 | - | 11 - 12 | 1101 | 94 | ||
L00619.1 | - | 11 - 12 | 1166 | 94 | ||
L05631.1 | - | 11 - 12 | 1127 | 94 | ||
L10628.1 | - | 11 - 12 | 1129 | 94 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
