Assay Details
Gene Symbol: Pdgfra | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001083316.1 | NP_001076785.1 | 2 - 3 | 242 | 75 | |
NM_011058.2 | NP_035188.2 | 2 - 3 | 225 | 75 | ||
GenBank mRNA | AK035501.1 | - | 2 - 3 | 182 | 75 | |
AK081664.1 | - | 2 - 3 | 224 | 75 | ||
AK136490.1 | - | 2 - 3 | 211 | 75 | ||
AK139094.1 | - | 1 - 2 | 457 | 75 | ||
AK147267.1 | - | 2 - 3 | 174 | 75 | ||
AK163952.1 | - | 2 - 3 | 289 | 75 | ||
BC020405.1 | - | 2 - 3 | 156 | 75 | ||
BC037467.1 | - | 2 - 3 | 164 | 75 | ||
BC053036.1 | - | 2 - 3 | 242 | 5704645 | 75 | |
CJ099401.1 | - | 2 - 3 | 225 | 75 | ||
M57683.1 | - | 2 - 3 | 124 | 75 | ||
M84607.1 | - | 2 - 3 | 235 | 75 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
