Assay Details
Gene Symbol: Pdgfra | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001083316.1 | NP_001076785.1 | 6 - 7 | 1128 | 84 | |
NM_011058.2 | NP_035188.2 | 6 - 7 | 1111 | 84 | ||
GenBank mRNA | AK035501.1 | - | 6 - 7 | 1068 | 84 | |
AK081664.1 | - | 6 - 7 | 1110 | 84 | ||
AK136490.1 | - | 6 - 7 | 1097 | 84 | ||
AK139094.1 | - | 5 - 6 | 1343 | 84 | ||
AK145097.1 | - | 6 - 7 | 1711 | 84 | ||
AK147267.1 | - | 6 - 7 | 1060 | 84 | ||
AK163952.1 | - | 6 - 7 | 1175 | 84 | ||
BC020405.1 | - | 6 - 7 | 1042 | 84 | ||
BC037467.1 | - | 6 - 7 | 1050 | 84 | ||
BC053036.1 | - | 6 - 7 | 1128 | 5704645 | 84 | |
M84607.1 | - | 6 - 7 | 1121 | 84 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
