Assay Details
Gene Symbol: Prkcd | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_011103.3 | NP_035233.1 | 15 - 16 | 1789 | 67 | |
GenBank mRNA | AB201453.1 | - | 12 - 13 | 1391 | 67 | |
AB201454.1 | - | 12 - 13 | 1469 | 67 | ||
AB201455.1 | - | 12 - 13 | 1391 | 67 | ||
AB201456.1 | - | 12 - 13 | 1469 | 67 | ||
AF251036.1 | - | 14 - 15 | 1552 | 67 | ||
AK144231.1 | - | 15 - 16 | 1794 | 67 | ||
AK168768.1 | - | 15 - 16 | 1784 | 67 | ||
AY545076.1 | - | 14 - 15 | 1552 | 67 | ||
HQ840432.1 | - | 7 - 8 | 643 | 67 | ||
X60304.1 | - | 14 - 15 | 1565 | 67 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
