Assay Details
Gene Symbol: Prkcd | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_011103.3 | NP_035233.1 | 16 - 17 | 1975 | 70 | |
GenBank mRNA | AB011812.1 | - | 15 - 16 | 1819 | 70 | |
AB201453.1 | - | 13 - 14 | 1577 | 70 | ||
AB201454.1 | - | 13 - 14 | 1655 | 70 | ||
AB201455.1 | - | 13 - 14 | 1577 | 70 | ||
AB201456.1 | - | 13 - 14 | 1655 | 70 | ||
AF251036.1 | - | 15 - 16 | 1738 | 70 | ||
AK144231.1 | - | 16 - 17 | 1980 | 70 | ||
AK168768.1 | - | 16 - 17 | 1970 | 70 | ||
AK171476.1 | - | 16 - 17 | 1980 | 70 | ||
AY545076.1 | - | 15 - 16 | 1738 | 70 | ||
HQ840432.1 | - | 8 - 9 | 829 | 70 | ||
M69042.1 | - | 16 - 17 | 1970 | 70 | ||
X60304.1 | - | 15 - 16 | 1751 | 70 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
