Assay Details
Gene Symbol: Syk | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001198977.1 | NP_001185906.1 | 6 - 7 | 1037 | 83 | |
NM_011518.2 | NP_035648.2 | 6 - 7 | 1041 | 83 | ||
GenBank mRNA | AK136363.1 | - | 6 - 7 | 1040 | 83 | |
AK143564.1 | - | 6 - 7 | 1070 | 83 | ||
BC065121.1 | - | 6 - 7 | 973 | 30533300 | 83 | |
U25685.1 | - | 5 - 6 | 825 | 83 | ||
U36776.1 | - | 6 - 7 | 1044 | 83 | ||
Z49877.1 | - | 6 - 7 | 1302 | 83 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
