Assay Details
Gene Symbol: Gria2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001039195.1 | NP_001034284.1 | 1 - 2 | 487 | 91 | |
NM_001083806.1 | NP_001077275.1 | 1 - 2 | 487 | 91 | ||
NM_013540.2 | NP_038568.2 | 1 - 2 | 487 | 91 | ||
GenBank mRNA | AB109210.1 | - | 1 - 2 | 80 | 91 | |
AB109211.1 | - | 1 - 2 | 80 | 91 | ||
AB111957.1 | - | 1 - 2 | 80 | 91 | ||
AK014389.1 | - | 1 - 2 | 532 | 91 | ||
AK043490.1 | - | 1 - 2 | 460 | 91 | ||
AK044574.1 | - | 1 - 2 | 419 | 91 | ||
AK045738.1 | - | 1 - 2 | 390 | 91 | ||
AK046861.1 | - | 1 - 2 | 487 | 91 | ||
AK086614.1 | - | 1 - 2 | 420 | 91 | ||
AK134572.1 | - | 1 - 2 | 418 | 91 | ||
AK141525.1 | - | 1 - 2 | 462 | 91 | ||
AK163941.1 | - | 1 - 2 | 480 | 91 | ||
BC048248.1 | - | 1 - 2 | 346 | 6494013 | 91 | |
BC066193.1 | - | 1 - 2 | 275 | 91 | ||
X57498.1 | - | 1 - 2 | 282 | 91 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
