Assay Details
Gene Symbol: Sema3c | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_013657.5 | NP_038685.3 | 11 - 12 | 1323 | 110 | |
GenBank mRNA | AK034239.1 | - | 11 - 12 | 1382 | 110 | |
BC066852.1 | - | 11 - 12 | 1323 | 30093230 | 110 | |
X85994.1 | - | 11 - 12 | 1275 | 110 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
