Assay Details
Gene Symbol: Ndrg3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_013865.2 | NP_038893.1 | 7 - 8 | 603 | 76 | |
NM_180956.1 | NP_851287.1 | 7 - 8 | 603 | 76 | ||
GenBank mRNA | AB033922.1 | - | 7 - 8 | 517 | 76 | |
AK002272.1 | - | 6 - 7 | 517 | 76 | ||
AK004689.1 | - | 7 - 8 | 549 | 76 | ||
AK036277.1 | - | 7 - 8 | 528 | 76 | ||
AK037487.1 | - | 7 - 8 | 529 | 76 | ||
AK154927.1 | - | 7 - 8 | 570 | 76 | ||
AK209466.1 | - | 4 - 5 | 317 | 76 | ||
AK214669.1 | - | 3 - 4 | 173 | 76 | ||
AK218281.1 | - | 3 - 4 | 173 | 76 | ||
BC018504.1 | - | 7 - 8 | 537 | 4500970 | 76 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
