Assay Details
Gene Symbol: Ndrg3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_013865.2 | NP_038893.1 | 12 - 13 | 969 | 130 | |
NM_180956.1 | NP_851287.1 | 12 - 13 | 969 | 130 | ||
GenBank mRNA | AB033922.1 | - | 12 - 13 | 883 | 130 | |
AK004689.1 | - | 12 - 13 | 915 | 130 | ||
AK037487.1 | - | 12 - 13 | 895 | 130 | ||
AK154927.1 | - | 12 - 13 | 936 | 130 | ||
AK185967.1 | - | 2 - 3 | 109 | 130 | ||
BC018504.1 | - | 12 - 13 | 903 | 4500970 | 130 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
