Assay Details
Gene Symbol: Mapk12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_013871.3 | NP_038899.1 | 1 - 2 | 358 | 51 | |
GenBank mRNA | AK165741.1 | - | 1 - 2 | 359 | 51 | |
BC021640.1 | - | 1 - 2 | 276 | 3979667 | 51 | |
BY733534.1 | - | 1 - 2 | 359 | 51 | ||
Y13439.1 | - | 1 - 2 | 294 | 51 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
