Assay Details
Gene Symbol: Arhgef7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001113517.1 | NP_001106989.1 | 10 - 11 | 1357 | 115 | |
NM_001113518.1 | NP_001106990.1 | 9 - 10 | 1146 | 115 | ||
NM_017402.4 | NP_059098.2 | 9 - 10 | 1146 | 115 | ||
GenBank mRNA | AF247654.1 | - | 9 - 10 | 1042 | 115 | |
AF247655.1 | - | 7 - 8 | 763 | 115 | ||
AF343877.1 | - | 9 - 10 | 1026 | 115 | ||
AK052545.1 | - | 9 - 10 | 1117 | 115 | ||
AK129064.1 | - | 10 - 11 | 1293 | 115 | ||
AK139156.1 | - | 10 - 11 | 1286 | 115 | ||
AY220301.1 | - | 9 - 10 | 1111 | 115 | ||
BC044838.1 | - | 9 - 10 | 1131 | 3499604 | 115 | |
U96634.1 | - | 7 - 8 | 836 | 115 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
