Assay Details
Gene Symbol: Gabbr1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_019439.3 | NP_062312.3 | 12 - 13 | 1891 | 59 | |
GenBank mRNA | AF008649.1 | - | 12 - 13 | 1775 | 59 | |
AF114168.2 | - | 12 - 13 | 1789 | 59 | ||
AF120255.1 | - | 7 - 8 | 1219 | 59 | ||
AK038844.1 | - | 931 | 59 | |||
AK156397.1 | - | 11 - 12 | 1776 | 59 | ||
BC054735.1 | - | 12 - 13 | 1891 | 6401243 | 59 | |
BC056990.1 | - | 7 - 8 | 1268 | 6821442 | 59 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
