Assay Details
Gene Symbol: Gria4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001113180.1 | NP_001106651.1 | 12 - 13 | 2326 | 76 | |
NM_019691.4 | NP_062665.3 | 12 - 13 | 2326 | 76 | ||
GenBank mRNA | AB022913.1 | - | 12 - 13 | 2060 | 76 | |
AB079073.1 | - | 11 - 12 | 1847 | 76 | ||
AB102777.1 | - | 11 - 12 | 1847 | 76 | ||
AB104708.1 | - | 1 - 2 | 211 | 76 | ||
AK031568.1 | - | 12 - 13 | 2300 | 76 | ||
AK036193.1 | - | 1 - 2 | 209 | 76 | ||
AK135034.1 | - | 12 - 13 | 2033 | 76 | ||
BC060697.1 | - | 12 - 13 | 2234 | 6409918 | 76 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
