Assay Details
Gene Symbol: Sec23b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001252543.1 | NP_001239472.1 | 4 - 5 | 482 | 80 | |
NM_001252544.1 | NP_001239473.1 | 4 - 5 | 475 | 80 | ||
NM_001252545.1 | NP_001239474.1 | 4 - 5 | 464 | 80 | ||
NM_019787.4 | NP_062761.2 | 4 - 5 | 493 | 80 | ||
GenBank mRNA | AF200326.1 | - | 3 - 4 | 365 | 80 | |
AK014597.1 | - | 4 - 5 | 443 | 80 | ||
AK139729.1 | - | 4 - 5 | 407 | 80 | ||
AK146083.1 | - | 4 - 5 | 442 | 80 | ||
AK149693.1 | - | 4 - 5 | 422 | 80 | ||
AK153214.1 | - | 4 - 5 | 449 | 80 | ||
AK155168.1 | - | 4 - 5 | 423 | 80 | ||
AK158398.1 | - | 4 - 5 | 460 | 80 | ||
AK166330.1 | - | 4 - 5 | 425 | 80 | ||
AK167382.1 | - | 4 - 5 | 431 | 80 | ||
AK167732.1 | - | 4 - 5 | 411 | 80 | ||
AK171597.1 | - | 4 - 5 | 423 | 80 | ||
BC005464.1 | - | 4 - 5 | 418 | 3591420 | 80 | |
BC011160.1 | - | 4 - 5 | 445 | 4024353 | 80 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
