Assay Details
Gene Symbol: Tk2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_021028.3 | NP_066356.3 | 1 - 2 | 159 | 112 | |
NR_045642.1 | 1 - 2 | 159 | 112 | |||
GenBank mRNA | AF105217.1 | - | 1 - 2 | 134 | 112 | |
AJ249341.2 | - | 1 - 2 | 137 | 112 | ||
AK041651.1 | - | 1 - 2 | 151 | 112 | ||
AK051286.1 | - | 1 - 2 | 153 | 112 | ||
AK087557.1 | - | 1 - 2 | 160 | 112 | ||
AK154160.1 | - | 1 - 2 | 153 | 112 | ||
BC019982.1 | - | 1 - 2 | 151 | 4159742 | 112 | |
BY083126.1 | - | 1 - 2 | 154 | 112 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
