Assay Details
Gene Symbol: Itgb6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001159564.1 | NP_001153036.1 | 6 - 7 | 686 | 69 | |
NM_021359.3 | NP_067334.1 | 3 - 4 | 580 | 69 | ||
GenBank mRNA | AF115376.1 | - | 6 - 7 | 532 | 69 | |
AK008867.1 | - | 3 - 4 | 581 | 69 | ||
AK019511.1 | - | 3 - 4 | 443 | 69 | ||
AK036439.1 | - | 6 - 7 | 687 | 69 | ||
AK085900.1 | - | 6 - 7 | 727 | 69 | ||
BC049185.1 | - | 3 - 4 | 549 | 6475423 | 69 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
