Assay Details
Gene Symbol: Cacna1h | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001163691.1 | NP_001157163.1 | 22 - 23 | 4716 | 78 | |
NM_021415.4 | NP_067390.4 | 22 - 23 | 4716 | 78 | ||
GenBank mRNA | AK122452.1 | - | 11 - 12 | 1689 | 78 | |
AK135951.1 | - | 4 - 5 | 463 | 78 | ||
AK147242.1 | - | 22 - 23 | 4717 | 78 | ||
BC138026.1 | - | 22 - 23 | 4555 | 8861045 | 78 | |
CA327685.1 | - | 5 - 6 | 537 | 78 | ||
CB246265.1 | - | 1 - 2 | 129 | 78 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
