Assay Details
Gene Symbol: Chrnd | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_021600.3 | NP_067611.2 | 2 - 3 | 301 | 82 | |
GenBank mRNA | AK081344.1 | - | 1 - 2 | 287 | 82 | |
BC052153.1 | - | 2 - 3 | 246 | 6336578 | 82 | |
BC141376.1 | - | 2 - 3 | 229 | 9055989 | 82 | |
BC141387.1 | - | 2 - 3 | 229 | 9056000 | 82 | |
K02582.1 | - | 2 - 3 | 212 | 82 | ||
L10076.1 | - | 2 - 3 | 261 | 82 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
