Assay Details
Gene Symbol: Chrnd | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_021600.3 | NP_067611.2 | 5 - 6 | 615 | 72 | |
GenBank mRNA | AK081344.1 | - | 4 - 5 | 601 | 72 | |
BC052153.1 | - | 5 - 6 | 560 | 6336578 | 72 | |
BC141376.1 | - | 5 - 6 | 543 | 9055989 | 72 | |
BC141387.1 | - | 5 - 6 | 543 | 9056000 | 72 | |
K02582.1 | - | 5 - 6 | 526 | 72 | ||
L10076.1 | - | 5 - 6 | 575 | 72 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
