Assay Details
Gene Symbol: Chrnd | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_021600.3 | NP_067611.2 | 8 - 9 | 1031 | 60 | |
GenBank mRNA | AK081344.1 | - | 7 - 8 | 1017 | 60 | |
BC052153.1 | - | 8 - 9 | 976 | 6336578 | 60 | |
BC141376.1 | - | 8 - 9 | 959 | 9055989 | 60 | |
BC141387.1 | - | 8 - 9 | 959 | 9056000 | 60 | |
K02582.1 | - | 8 - 9 | 942 | 60 | ||
L10076.1 | - | 8 - 9 | 991 | 60 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
