Assay Details
Gene Symbol: Epha1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_023580.4 | NP_076069.2 | 12 - 13 | 2147 | 98 | |
GenBank mRNA | AF131197.1 | - | 12 - 13 | 2089 | 98 | |
AK028478.1 | - | 12 - 13 | 2148 | 98 | ||
BC071215.1 | - | 12 - 13 | 2111 | 4196138 | 98 | |
U18084.1 | - | 8 - 9 | 1250 | 98 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
