Assay Details
Gene Symbol: Dmpk | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001190490.1 | NP_001177419.1 | 3 - 4 | 483 | 105 | |
NM_001190491.1 | NP_001177420.1 | 3 - 4 | 483 | 105 | ||
NM_032418.2 | NP_115794.1 | 3 - 4 | 483 | 105 | ||
GenBank mRNA | BC024150.1 | - | 3 - 4 | 470 | 4217643 | 105 |
BC056615.1 | - | 3 - 4 | 462 | 105 | ||
BC075715.1 | - | 3 - 4 | 391 | 4989259 | 105 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
