Assay Details
Gene Symbol: Dmpk | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001190490.1 | NP_001177419.1 | 11 - 12 | 1416 | 85 | |
NM_001190491.1 | NP_001177420.1 | 10 - 11 | 1494 | 85 | ||
NM_032418.2 | NP_115794.1 | 10 - 11 | 1494 | 85 | ||
GenBank mRNA | BC024150.1 | - | 10 - 11 | 1481 | 4217643 | 85 |
BC056615.1 | - | 10 - 11 | 1458 | 85 | ||
BC075715.1 | - | 11 - 12 | 1324 | 4989259 | 85 | |
S60313.1 | - | 7 - 8 | 85 | |||
S60314.1 | - | 7 - 8 | 85 | |||
S60315.1 | - | 8 - 9 | 85 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
