Assay Details
Gene Symbol: Prkcsh | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001293650.1 | NP_001280579.1 | 17 - 18 | 1752 | 79 | |
NM_001293651.1 | NP_001280580.1 | 17 - 18 | 1735 | 79 | ||
NM_008925.2 | NP_032951.1 | 17 - 18 | 1731 | 79 | ||
GenBank mRNA | AK137661.1 | - | 17 - 18 | 1698 | 79 | |
AK153931.1 | - | 17 - 18 | 1706 | 79 | ||
AK159705.1 | - | 17 - 18 | 1702 | 79 | ||
AK204280.1 | - | 1 - 2 | 48 | 79 | ||
BC009816.1 | - | 17 - 18 | 1685 | 4008182 | 79 | |
U92794.1 | - | 17 - 18 | 1684 | 79 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
in utero embryonic development
liver development
N-glycan processing
nitrogen compound metabolic process
negative regulation of neuron projection development
protein heterooligomerization
renal system development
RNA binding
protein kinase C binding
calcium ion binding
protein binding
ion channel binding
metal ion binding
phosphoprotein binding
in utero embryonic development
liver development
N-glycan processing
nitrogen compound metabolic process
negative regulation of neuron projection development
protein heterooligomerization
renal system development
RNA binding
protein kinase C binding
calcium ion binding
protein binding
ion channel binding
metal ion binding
phosphoprotein binding
MicroArray Probes:
Agilent
Affymetrix
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P26349967 | - | - |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2035892 | BC009816.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2035892 | U92794.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2035892 | AK153931.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2035892 | AK159705.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2035892 | NM_008925.1 | mouse_transcriptome |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 176706 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 331516 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 185482 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 627891 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 839368 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 85680 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 725359 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 126610 | - | - |
Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P135678 | - | - |
Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P115159 | - | - |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2035892 | BC009816.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2035892 | U92794.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2035892 | AK153931.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2035892 | AK159705.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2035892 | NM_008925.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P520690 | BC009816.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P520690 | U92794.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P520690 | AK153931.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P520690 | AK159705.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P520690 | AK204280.1 | mouse_transcriptome |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26349968 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26349997 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26349986 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26349993 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26349978 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26349990 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26349976 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26349975 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26349972 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26349994 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26349970 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26349984 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26349981 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26349988 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26349979 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26349987 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26349973 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26349999 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26349971 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26349974 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P115159 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P135678 | - | - |
Whole Mouse Genome G4122A | A_51_P520690 | BC009816.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P520690 | U92794.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P520690 | AK153931.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P520690 | AK159705.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P520690 | AK204280.1 | mouse_transcriptome |
Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P108805 | - | - |
Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P135678 | - | - |
Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P115159 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P07653994 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P07653999 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P26349967 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26349970 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26350018 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26350037 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26349988 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26350027 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26349966 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26349965 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26350005 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26350044 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26349987 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26350054 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26350023 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26349993 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26349981 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26350056 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26349997 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26350028 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26350032 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26350026 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26350060 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26350020 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26350029 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26349975 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26350052 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26349982 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26349979 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26350016 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26350004 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26350022 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26349990 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26350001 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26350038 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26350006 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26350035 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26349972 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26350058 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26349974 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26350014 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26350013 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26350057 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26350033 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26349986 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26349968 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26349980 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26350051 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26350047 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26350025 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26349964 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26349984 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26350040 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26350052 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26350054 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26349990 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26349966 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P07823313 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P07823322 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P08137901 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26350006 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26350060 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26349982 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26349978 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26350056 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26350016 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P07690752 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P07985671 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26350014 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26350035 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26350047 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P07640305 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P08137904 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26350025 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26350038 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26350001 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26350042 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26350010 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26349970 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26349976 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26350058 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26349981 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26349979 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26349968 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P07690751 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P08137910 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26350044 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26349997 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P08123782 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P08198266 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P08123774 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26350040 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26350037 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26349993 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26350005 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P08198261 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26349965 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26350023 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26350004 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26349995 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26349977 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P07654003 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26350013 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P07654010 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P07823316 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26349971 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26349980 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26350020 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Mouse Gene 1.0 ST Array | 10583797 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10583804 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10583805 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10583790 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10583799 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10583798 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10583789 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10583792 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10583801 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10583791 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10583796 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10583794 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10583800 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10583793 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10583802 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10583803 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10583795 | - | - |
Affymetrix HT MG-430 PM Array Plate | 1416339_PM_a_at | - | - |
Affymetrix Mouse Gene 1.1 ST Array | 10583788 | - | - |
Mouse Genome 430 2.0 Array | 1416339_PM_a_at | - | - |
Mouse Genome 430 2.0 Array | 1416339_a_at | - | - |
Affymetrix GeneChip Mouse Genome 430 2.0 Array | 19089_at | - | - |
Affymetrix GeneChip Mouse Genome 430 2.0 Array | 19089_PM_at | - | - |
Affymetrix Mouse Clariom S Array | PSR0900001613.mm.1 | NM_008925.2 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0900001613.mm.1 | TC0900000210.mm.1 | MTA TC |
Mouse Expression Set 430 | 1416339_a_at | - | - |
Mouse Expression Set 430 | 1416339_PM_a_at | - | - |
Murine Genome U74v2 Set | 100684_at | - | - |
Murine Genome U74v2 Set | 100684_PM_at | - | - |
Mouse Genome 430A 2.0 Array | 1416339_PM_a_at | - | - |
Mouse Genome 430A 2.0 Array | 1416339_a_at | - | - |
Affymetrix HT Mouse Genome 430A Array | 1416339_PM_a_at | - | - |
Affymetrix HT Mouse Genome 430A Array | 1416339_a_at | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
MouseWG-6 v2 Expression BeadChip | 3360398 | - | - |
Illumina mouse-6 v1.0 expression beadchip | 3360398 | - | - |
Illumina mouse-6 v1.0 expression beadchip | 105420541 | - | - |
Illumina mouseRef-8 v1.0 expression beadchip | 0003360398 | - | - |
Illumina mouseRef-8 v1.1 expression beadchip | ILMN_1233076 | - | - |
Illumina mouse-6 v1.1 expression beadchip | ILMN_1255077 | - | - |
Illumina mouse-6 v1.1 expression beadchip | ILMN_1233076 | - | - |
mouse-6 v1.1 (Illumina) | scl37209.17.10_83-S | - | - |
Illumina Sentrix mouse-6.v1 bead array | 105420541 | - | - |
Illumina Sentrix mouse-6.v1 bead array | 3360398 | - | - |
Illumina Sentrix MouseRef-8 Expression BeadChip | scl37209.17.10_83-S | - | - |
Illumina Sentrix Mouse-6 Expression BeadChip (2) | 3360398 | - | - |
Illumina Sentrix Mouse-6 Expression BeadChip (2) | 105420541 | - | - |
MouseRef-8 v2 Expression BeadChip | ILMN_1233076 | - | - |
Illumina MouseWG-6 v2.0 expression beadchip | ILMN_1233076 | - | - |
Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2555646 | - | - |
Illumina Sentrix Mouse-6 Expression BeadChip (3) | scl37209.17.10_83-S | - | - |
