Assay Details
Gene Symbol: Stat6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_009284.2 | NP_033310.2 | 20 - 21 | 2486 | 88 | |
GenBank mRNA | AF481808.1 | - | 19 - 20 | 2201 | 88 | |
AF481809.1 | - | 19 - 20 | 2201 | 88 | ||
AK135911.1 | - | 3 - 4 | 248 | 88 | ||
AK149563.1 | - | 20 - 21 | 2447 | 88 | ||
AK163053.1 | - | 20 - 21 | 2487 | 88 | ||
BC029318.1 | - | 7 - 8 | 675 | 88 | ||
BC127054.1 | - | 20 - 21 | 2276 | 40109712 | 88 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
