Assay Details
Gene Symbol: Musk | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001037127.2 | NP_001032204.2 | 10 - 11 | 1407 | 74 | |
NM_001037128.1 | NP_001032205.1 | 10 - 11 | 1362 | 74 | ||
NM_001037129.1 | NP_001032206.1 | 10 - 11 | 1362 | 74 | ||
NM_001037130.1 | NP_001032207.1 | 9 - 10 | 1332 | 74 | ||
NM_001165996.1 | NP_001159468.1 | 9 - 10 | 1332 | 74 | ||
NM_010944.2 | NP_035074.2 | 9 - 10 | 1332 | 74 | ||
GenBank mRNA | AB691772.1 | - | 9 - 10 | 1182 | 74 | |
AY360453.1 | - | 9 - 10 | 1227 | 74 | ||
AY955296.1 | - | 10 - 11 | 1257 | 74 | ||
AY955297.1 | - | 10 - 11 | 1212 | 74 | ||
AY955298.1 | - | 10 - 11 | 1212 | 74 | ||
AY956403.1 | - | 9 - 10 | 1182 | 74 | ||
AY956404.1 | - | 9 - 10 | 1182 | 74 | ||
BC100342.1 | - | 10 - 11 | 1286 | 30935290 | 74 | |
U37708.1 | - | 9 - 10 | 1330 | 74 | ||
U37709.1 | - | 9 - 10 | 1330 | 74 | ||
X86444.1 | - | 8 - 9 | 1236 | 74 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
