Assay Details
Gene Symbol: Pfdn2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_011070.3 | NP_035200.2 | 2 - 3 | 176 | 70 | |
GenBank mRNA | BC026839.1 | - | 2 - 3 | 163 | 4219635 | 70 |
BC049606.1 | - | 2 - 3 | 176 | 6535749 | 70 | |
Y17393.1 | - | 2 - 3 | 166 | 70 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
