Assay Details
Gene Symbol: Pfdn2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_011070.3 | NP_035200.2 | 3 - 4 | 300 | 63 | |
GenBank mRNA | BC026839.1 | - | 3 - 4 | 287 | 4219635 | 63 |
BC049606.1 | - | 3 - 4 | 300 | 6535749 | 63 | |
Y17393.1 | - | 3 - 4 | 290 | 63 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P20827418 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P20827419 | - | - |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P327778 | BC049606.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P327778 | Y17393.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P327778 | BC026839.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P327778 | NM_011070.3 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P327778 | BC049606.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P327778 | Y17393.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P327778 | BC026839.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P327778 | NM_011070.3 | mouse_transcriptome |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 623966 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 494332 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 144698 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 402721 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 756462 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 941013 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 87643 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 113268 | - | - |
Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P120384 | - | - |
Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P139909 | - | - |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P327778 | BC049606.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P327778 | Y17393.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P327778 | BC026839.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P327778 | NM_011070.3 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P327778 | BC049606.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P327778 | Y17393.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P327778 | BC026839.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P327778 | NM_011070.3 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P327778 | BC049606.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P327778 | Y17393.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P327778 | BC026839.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P327778 | NM_011070.3 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P327778 | BC049606.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P327778 | Y17393.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P327778 | BC026839.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P327778 | NM_011070.3 | mouse_transcriptome |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20827434 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20827440 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20827418 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20827426 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20827428 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20827429 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20827422 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20827417 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20827432 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20827436 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20827438 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20827424 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20827431 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P120384 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P04354143 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P139909 | - | - |
Whole Mouse Genome G4122A | A_51_P327778 | BC049606.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P327778 | Y17393.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P327778 | BC026839.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P327778 | NM_011070.3 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P327778 | BC049606.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P327778 | Y17393.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P327778 | BC026839.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P327778 | NM_011070.3 | mouse_transcriptome |
Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P136573 | - | - |
Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P139909 | - | - |
Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P120384 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P00516293 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P00516306 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P20827418 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P20827419 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20827507 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20827516 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20827522 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20827482 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20827428 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20827437 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20827490 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20827518 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20827426 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20827429 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20827526 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20827458 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20827472 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20827486 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20827417 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20827452 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20827470 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20827431 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20827520 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20827531 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20827491 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20827494 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20827497 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20827519 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20827523 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20827527 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20827503 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20827447 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20827524 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20827529 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20827479 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20827501 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20827480 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20827514 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20827425 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20827440 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20827445 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20827477 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20827488 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20827436 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20827418 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20827484 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20827460 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20827530 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20827422 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20827508 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20827466 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20827475 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20827495 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20827510 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20827532 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20827442 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20827462 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20827513 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20827528 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20827431 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P01055700 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P01055699 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20827453 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20827430 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20827424 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P01055697 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20827433 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20827455 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P01070003 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P00394901 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20827449 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P00603440 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P01055696 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20827450 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20827425 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20827418 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20827454 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P01055698 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P01070004 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20827428 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20827434 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20827436 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20827439 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20827440 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P01070006 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20827460 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P00394900 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20827426 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P00560791 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20827451 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20827422 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P01085624 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P00650992 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Mouse Gene 1.0 ST Array | 10351587 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10351586 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10351584 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10351585 | - | - |
Affymetrix HT MG-430 PM Array Plate | 1421950_PM_at | - | - |
Affymetrix Mouse Gene 1.1 ST Array | 10351583 | - | - |
Mouse Genome 430 2.0 Array | 1421950_at | - | - |
Mouse Genome 430 2.0 Array | 1421950_PM_at | - | - |
Affymetrix GeneChip Mouse Genome 430 2.0 Array | 18637_PM_at | - | - |
Affymetrix GeneChip Mouse Genome 430 2.0 Array | 18637_at | - | - |
Affymetrix Mouse Clariom S Array | PSR0100013060.mm.1 | TC0100001586.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR0100013060.mm.1 | NM_011070.3.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0100013060.mm.1 | NM_011070.3 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0100013059.mm.1 | TC0100001586.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR0100013059.mm.1 | NM_011070.3.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0100013059.mm.1 | NM_011070.3 | Refseq |
Mouse Expression Set 430 | 1421950_at | - | - |
Mouse Expression Set 430 | 1421950_PM_at | - | - |
Murine Genome U74v2 Set | 102252_at | - | - |
Murine Genome U74v2 Set | 102252_PM_at | - | - |
Mouse Genome 430A 2.0 Array | 1421950_at | - | - |
Mouse Genome 430A 2.0 Array | 1421950_PM_at | - | - |
Affymetrix HT Mouse Genome 430A Array | 1421950_PM_at | - | - |
Affymetrix HT Mouse Genome 430A Array | 1421950_at | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Mouse Genome Survey Microarray | 906745 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Illumina mouse-6 v1.0 expression beadchip | 4230156 | - | - |
Illumina mouseRef-8 v1.0 expression beadchip | 0004230156 | - | - |
Illumina mouseRef-8 v1.1 expression beadchip | ILMN_1219667 | - | - |
Illumina mouse-6 v1.1 expression beadchip | ILMN_1219667 | - | - |
mouse-6 v1.1 (Illumina) | scl018637.3_29-S | - | - |
Illumina Sentrix mouse-6.v1 bead array | 4230156 | - | - |
Illumina Sentrix MouseRef-8 Expression BeadChip | scl018637.3_29-S | - | - |
Illumina Sentrix Mouse-6 Expression BeadChip (2) | 4230156 | - | - |
MouseRef-8 v2 Expression BeadChip | ILMN_1219667 | - | - |
Illumina MouseWG-6 v2.0 expression beadchip | ILMN_1219667 | - | - |
Illumina Sentrix Mouse-6 Expression BeadChip (3) | scl018637.3_29-S | - | - |
