Assay Details
Gene Symbol: Ptpre | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_011212.3 | NP_035342.3 | 16 - 17 | 1784 | 79 | |
GenBank mRNA | AK004602.1 | - | 16 - 17 | 1786 | 79 | |
AK132455.1 | - | 15 - 16 | 1747 | 79 | ||
AK154910.1 | - | 13 - 14 | 1336 | 79 | ||
BC053408.1 | - | 16 - 17 | 1560 | 79 | ||
BC098473.1 | - | 16 - 17 | 1548 | 79 | ||
D83484.1 | - | 16 - 17 | 2121 | 79 | ||
U35368.1 | - | 16 - 17 | 1784 | 79 | ||
U36758.1 | - | 13 - 14 | 1333 | 79 | ||
U40280.1 | - | 15 - 16 | 1524 | 79 | ||
U62387.1 | - | 16 - 17 | 1686 | 79 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
