Assay Details
Gene Symbol: Nbn | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_013752.3 | NP_038780.3 | 13 - 14 | 2176 | 109 | |
GenBank mRNA | AB016988.1 | - | 13 - 14 | 2098 | 109 | |
AF076687.1 | - | 13 - 14 | 2170 | 109 | ||
AF092840.1 | - | 13 - 14 | 2159 | 109 | ||
AK132302.1 | - | 13 - 14 | 2218 | 109 | ||
AK159790.1 | - | 13 - 14 | 2182 | 109 | ||
AK160614.1 | - | 13 - 14 | 2143 | 109 | ||
BC003719.1 | - | 13 - 14 | 2185 | 109 | ||
BC055061.1 | - | 13 - 14 | 2168 | 6528890 | 109 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
