Assay Details
Gene Symbol: Sh2d3c | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001252547.1 | NP_001239476.1 | 3 - 4 | 284 | 64 | |
NM_013781.3 | NP_038809.1 | 4 - 5 | 770 | 64 | ||
NR_045535.1 | 5 - 6 | 1228 | 64 | |||
GenBank mRNA | AB030442.1 | - | 3 - 4 | 247 | 64 | |
AB043953.1 | - | 4 - 5 | 753 | 64 | ||
AF168364.1 | - | 4 - 5 | 711 | 64 | ||
AK042709.1 | - | 3 - 4 | 268 | 64 | ||
AK085885.1 | - | 269 | 64 | |||
AK155165.1 | - | 4 - 5 | 706 | 64 | ||
AK172072.1 | - | 5 - 6 | 1149 | 64 | ||
BC113203.1 | - | 4 - 5 | 725 | 40048064 | 64 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
