Assay Details
Gene Symbol: Usp21 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_013919.4 | NP_038947.2 | 5 - 6 | 1113 | 73 | |
GenBank mRNA | AF177759.1 | - | 4 - 5 | 912 | 73 | |
AK011148.1 | - | 5 - 6 | 1123 | 73 | ||
AK013252.1 | - | 5 - 6 | 1108 | 73 | ||
BC021903.1 | - | 5 - 6 | 1103 | 3985792 | 73 | |
KF646669.1 | - | 4 - 5 | 631 | 73 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
