Assay Details
Gene Symbol: Usp21 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_013919.4 | NP_038947.2 | 11 - 12 | 1720 | 80 | |
GenBank mRNA | AF177759.1 | - | 10 - 11 | 1519 | 80 | |
AK011148.1 | - | 11 - 12 | 1730 | 80 | ||
BC021903.1 | - | 11 - 12 | 1710 | 3985792 | 80 | |
KF646669.1 | - | 10 - 11 | 1238 | 80 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
