Assay Details
Gene Symbol: Pde1a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001009978.1 | NP_001009978.1 | 6 - 7 | 658 | 70 | |
NM_001009979.1 | NP_001009979.1 | 6 - 7 | 658 | 70 | ||
NM_001159582.1 | NP_001153054.1 | 8 - 9 | 1160 | 70 | ||
NM_016744.4 | NP_058024.2 | 7 - 8 | 955 | 70 | ||
GenBank mRNA | AF159298.1 | - | 6 - 7 | 658 | 70 | |
AK043647.1 | - | 7 - 8 | 956 | 70 | ||
AY845863.1 | - | 7 - 8 | 707 | 70 | ||
AY845864.1 | - | 6 - 7 | 658 | 70 | ||
BC090628.1 | - | 7 - 8 | 923 | 6842675 | 70 | |
U56649.1 | - | 8 - 9 | 891 | 70 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
