Assay Details
Gene Symbol: Porcn | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AB036746.1 | - | 2 - 3 | 313 | 56 | |
AB036747.1 | - | 2 - 3 | 313 | 56 | ||
AB036748.1 | - | 2 - 3 | 313 | 56 | ||
AB036749.1 | - | 2 - 3 | 313 | 56 | ||
BC032284.1 | - | 2 - 3 | 283 | 5362485 | 56 | |
BF462782.1 | - | 2 - 3 | 306 | 56 | ||
BQ556448.1 | - | 1 - 2 | 130 | 56 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
MicroArray Probes:
