Assay Details
Gene Symbol: Polg | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_017462.2 | NP_059490.2 | 1 - 2 | 202 | 65 | |
GenBank mRNA | AK133584.1 | - | 1 - 2 | 128 | 65 | |
AK148091.1 | - | 1 - 2 | 164 | 65 | ||
BC053024.1 | - | 1 - 2 | 130 | 65 | ||
BC061472.1 | - | 1 - 2 | 116 | 6827238 | 65 | |
BC080291.1 | - | 1 - 2 | 109 | 65 | ||
CJ182989.1 | - | 1 - 2 | 203 | 65 | ||
U53584.1 | - | 1 - 2 | 122 | 65 |

More Information
Updated Mapping Information

Additional Information:
This assay is in the 5'UTR region of the AK133584.1,AK148091.1,BC061472.1,NM_017462.2,U53584.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
