Assay Details
Gene Symbol: Polg | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_017462.2 | NP_059490.2 | 21 - 22 | 3782 | 68 | |
GenBank mRNA | AB121698.1 | - | 20 - 21 | 3475 | 68 | |
AK133584.1 | - | 21 - 22 | 3708 | 68 | ||
AK148091.1 | - | 21 - 22 | 3744 | 68 | ||
AK162462.1 | - | 6 - 7 | 1351 | 68 | ||
BC053024.1 | - | 21 - 22 | 3708 | 68 | ||
BC061472.1 | - | 20 - 21 | 3774 | 6827238 | 68 | |
BC080291.1 | - | 21 - 22 | 3613 | 68 | ||
U53584.1 | - | 21 - 22 | 3702 | 68 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
